Tisk…

2-tert-Butylpyrazine 95%

Dodavatel: Apollo Scientific

OR920095-1G OR920095-5G
APOSOR920095-1GEA 1350 CZK
APOSOR920095-1G APOSOR920095-5G
2-tert-Butylpyrazine 95%
2-tert-Butylpyrazine
Vzorec: C₈H₁₂N₂
Storage Temperature: Ambient
MDL Number: MFCD00069193
CAS číslo: 32741-11-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR