(S)-5-Allyl-2-oxabicyclo[3.3.0]oct-8-ene 95%
Dodavatel: Apollo Scientific
OR920063-1G
OR920063-250MG
APOSOR920063-1GEA
5590
CZK
APOSOR920063-1G
APOSOR920063-250MG
(S)-5-Allyl-2-oxabicyclo[3.3.0]oct-8-ene 95%
(S)-5-Allyl-2-oxabicyclo[3.3.0]oct-8-ene
Vzorec:
C₁₀H₁₄O Storage Temperature: Ambient |
CAS číslo:
1052236-86-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...