Tisk…

2-Hydroxy-4-methoxyaniline hydrochloride 97%

Dodavatel: Apollo Scientific
Varování

OR919803-1G OR919803-250MG
APOSOR919803-1GEA 2400 CZK
APOSOR919803-1G APOSOR919803-250MG
2-Hydroxy-4-methoxyaniline hydrochloride 97%
2-Hydroxy-4-methoxyaniline hydrochloride
Vzorec: C₇H₉NO₂
Storage Temperature: Ambient
MDL Number: MFCD00052002
CAS číslo: 39547-15-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR