2,6-Dimethylcyclohexanol (mixture of isomers) 95%
Dodavatel: Apollo Scientific
Synonyms:
2,6-Dimethylcyclohexanol
OR919777-25G
OR919777-5G
APOSOR919777-25GEA
6020
CZK
APOSOR919777-25G
APOSOR919777-5G
2,6-Dimethylcyclohexanol (mixture of isomers) 95%
2,6-Dimethylcyclohexanol (mixture of isomers)
Vzorec:
C₈H₁₆O Storage Temperature: Ambient |
MDL Number:
MFCD00001502 CAS číslo: 5337-72-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...