2-Ethoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%
Dodavatel: Apollo Scientific
Synonyms:
Ethoxyboronic Acid Pinacol Ester
OR919768-25G
APOSOR919768-25GEA
6550
CZK
APOSOR919768-25G
2-Ethoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%
2-Ethoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Vzorec:
C₈H₁₇BO₃ MW: 172,03 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22054335 CAS číslo: 1126-93-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...