N,N-Diethylglycine methyl ester 95%
Dodavatel: Apollo Scientific
OR919729-100G
OR919729-25G
OR919729-5G
APOSOR919729-100GEA
20270
CZK
APOSOR919729-100G
APOSOR919729-25G
APOSOR919729-5G
N,N-Diethylglycine methyl ester 95%
N,N-Diethylglycine methyl ester
Vzorec:
C₇H₁₅NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD00059368 CAS číslo: 30280-35-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...