Tisk…

1-(2-Cyanoethyl)-2-methylpiperidine 95%

Dodavatel: Apollo Scientific

Synonyms: 1-(2-Cyanoethyl)-2-pipecoline

OR919695-25G OR919695-5G
APOSOR919695-25GEA 6220 CZK
APOSOR919695-25G APOSOR919695-5G
1-(2-Cyanoethyl)-2-methylpiperidine 95%
1-(2-Cyanoethyl)-2-methylpiperidine
Vzorec: C₉H₁₆N₂
MW: 152,24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00191397
CAS číslo: 140837-33-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR