Tisk…

1-Phenyl-2-butanone 95%

Dodavatel: Apollo Scientific

OR919635-1G OR919635-5G
APOSOR919635-1GEA 939 CZK
APOSOR919635-1G APOSOR919635-5G
1-Phenyl-2-butanone 95%
1-Phenyl-2-butanone
Vzorec: C₆H₅C(O)CH₂CH₂CH₃
Storage Temperature: Ambient
MDL Number: MFCD00009315
CAS číslo: 1007-32-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR