Tisk…

Valeramide 95%

Dodavatel: Apollo Scientific

Synonyms: Pentanamide

OR919589-25G OR919589-100G
APOSOR919589-25GEA 2690 CZK
APOSOR919589-25G APOSOR919589-100G
Valeramide 95%
Valeramide
Vzorec: C₅H₁₀O₂
Storage Temperature: Ambient
MDL Number: MFCD00041895
CAS číslo: 626-97-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR