Tisk…

2,3-Dimethoxybenzyl alcohol 95%

Dodavatel: Apollo Scientific

Synonyms: (2,3-Dimethoxyphenyl)methanol

OR919570-25G
APOSOR919570-25GEA 3230 CZK
APOSOR919570-25G
2,3-Dimethoxybenzyl alcohol 95%
2,3-Dimethoxybenzyl alcohol
Vzorec: C₉H₁₂O₃
MW: 168,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00004612
CAS číslo: 5653-67-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR