Tisk…

2-(Methylsulphonyl)acetophenone 95%

Dodavatel: Apollo Scientific

OR919426-10G OR919426-1G OR919426-5G
APOSOR919426-10GEA 25630 CZK
APOSOR919426-10G APOSOR919426-1G APOSOR919426-5G
2-(Methylsulphonyl)acetophenone 95%
2-(Methylsulphonyl)acetophenone
Vzorec: C₉H₁₀O₃S
Storage Temperature: Ambient
MDL Number: MFCD00025078
CAS číslo: 3708-04-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR