2,5-Dichloroterephthalonitrile 97%
Dodavatel: Apollo Scientific
OR919416-1G
OR919416-5G
APOSOR919416-1GEA
0
CZK
APOSOR919416-1G
APOSOR919416-5G
2,5-Dichloroterephthalonitrile 97%
2,5-Dichloroterephthalonitrile
Vzorec:
C₈H₂Cl₂N₂ Storage Temperature: Ambient |
MDL Number:
MFCD00059584 CAS číslo: 1897-43-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...