Tisk…

ɑ-Methylene-γ-valerolactone 97%

Dodavatel: Apollo Scientific

Synonyms: 4,5-Dihydro-5-methyl-3-methylene-2(3H)-furanone

OR919340-10G OR919340-1G OR919340-5G OR919340-25G
APOSOR919340-10GEA 0 CZK
APOSOR919340-10G APOSOR919340-1G APOSOR919340-5G APOSOR919340-25G
ɑ-Methylene-γ-valerolactone 97%
alpha-Methylene-gamma-valerolactone
Vzorec: C₆H₈O₂
MW: 112,13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06797118
CAS číslo: 62873-16-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR