4-(Bromomethyl)benzaldehyde 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR9193-1G
OR9193-250MG
OR9193-5G
APOSOR9193-1GEA
705
CZK
APOSOR9193-1G
APOSOR9193-250MG
APOSOR9193-5G
4-(Bromomethyl)benzaldehyde 97%
4-(Bromomethyl)benzaldehyde
Vzorec:
C₈H₇BrO Bod tání: 98,5…100 °C Storage Temperature: Ambient |
MDL Number:
MFCD08271963 CAS číslo: 51359-78-5 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...