Tisk…

Isopropyl methacrylate 95%

Dodavatel: Apollo Scientific

Synonyms: Methacrylic Acid Isopropyl Ester

OR919193-100G OR919193-25G
APOSOR919193-100GEA 7280 CZK
APOSOR919193-100G APOSOR919193-25G
Isopropyl methacrylate 95%
Isopropyl methacrylate
Vzorec: C₇H₁₂O₂
MW: 128.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00048290
CAS číslo: 4655-34-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR