Allyl cinnamate 98%
Dodavatel: Apollo Scientific
Synonyms:
Allyl Cinnamate, Cinnamic Acid Allyl Ester, Cinnamic acid Allyl Ester
OR919186-100G
OR919186-1KG
OR919186-500G
APOSOR919186-100GEA
3540
CZK
APOSOR919186-100G
APOSOR919186-1KG
APOSOR919186-500G
Allyl cinnamate 98%
Allyl cinnamate
Vzorec:
C₁₂H₁₂O₂ MW: 188,23 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00026105 CAS číslo: 1866-31-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...