Tisk…

1-Methylazepan-4-amine 96%

Dodavatel: Apollo Scientific

OR919163-1G OR919163-250MG
APOSOR919163-1GEA 25000 CZK
APOSOR919163-1G APOSOR919163-250MG
1-Methylazepan-4-amine 96%
1-Methylazepan-4-amine
Vzorec: C₇H₁₆N₂
Storage Temperature: Ambient
MDL Number: MFCD17015865
CAS číslo: 933741-93-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR