6-Methyl-2-oxo-1,2-dihydro-3-quinolinecarbaldehyde 95%
Dodavatel: Apollo Scientific
Synonyms:
6-Methyl-2-oxo-1,2-dihydroquinoline-3-carbaldehyde, 2-Hydroxy-6-methylquinoline-3-carbaldehyde
OR919139-1G
OR919139-250MG
APOSOR919139-1GEA
4600
CZK
APOSOR919139-1G
APOSOR919139-250MG
6-Methyl-2-oxo-1,2-dihydro-3-quinolinecarbaldehyde 95%
6-Methyl-2-oxo-1,2-dihydro-3-quinolinecarbaldehyde
Vzorec:
C₁₁H₉NO₂ MW: 187,2 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02227050 CAS číslo: 101382-53-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...