N-tert-Butyl-3,5-dimethylaniline 98%
Dodavatel: Apollo Scientific
OR918607-1G
OR918607-250MG
OR918607-5G
APOSOR918607-1GEA
5790
CZK
APOSOR918607-1G
APOSOR918607-250MG
APOSOR918607-5G
N-tert-Butyl-3,5-dimethylaniline 98%
N-tert-Butyl-3,5-dimethylaniline
Vzorec:
C₁₂H₁₉N Storage Temperature: Ambient |
MDL Number:
MFCD07440135 CAS číslo: 110993-40-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...