4-Methoxy-2,2,6,6-tetramethylpiperidinooxy (4-methoxy-TEMPO) 95%
Dodavatel: Apollo Scientific
Synonyms:
4-Methoxy-2,2,6,6-tetramethylpiperidine 1-Oxyl, 4-Methoxy-2,2,6,6-tetramethyl-1-piperidinyloxy
OR918603-1G
OR918603-5G
APOSOR918603-1GEA
1780
CZK
APOSOR918603-1G
APOSOR918603-5G
4-Methoxy-2,2,6,6-tetramethylpiperidinooxy (4-methoxy-TEMPO) 95%
4-Methoxy-2,2,6,6-tetramethylpiperidinooxy (4-methoxy-TEMPO)
Vzorec:
C₁₀H₂₀NO₂ MW: 186,27 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00270334 CAS číslo: 95407-69-5 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...