Tisk…

3-(Chloromethyl)-1-methyl-1H-1,2,4-triazole hydrochloride 95%

Dodavatel: Apollo Scientific

OR918236-1G OR918236-250MG OR918236-5G
APOSOR918236-1GEA 2200 CZK
APOSOR918236-1G APOSOR918236-250MG APOSOR918236-5G
3-(Chloromethyl)-1-methyl-1H-1,2,4-triazole hydrochloride 95%
3-(Chloromethyl)-1-methyl-1H-1,2,4-triazole hydrochloride
Vzorec: C₄H₇Cl₂N₃
Storage Temperature: Ambient
MDL Number: MFCD09701417
CAS číslo: 135206-76-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR