Tisk…

1,2:5,6-Diepoxyhexane 95%

Dodavatel: Apollo Scientific

Synonyms: 1,5-Hexadiene diepoxide

OR918172-5G
APOSOR918172-5GEA 2320 CZK
APOSOR918172-5G
1,2:5,6-Diepoxyhexane 95%
1,2:5,6-Diepoxyhexane
Vzorec: C₆H₁₀O₂
MW: 114,1424 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01723447
CAS číslo: 1888-89-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR