Tisk…

3-Ethyl-3-oxetanecarboxaldehyde 95%

Dodavatel: Apollo Scientific

OR918025-1G OR918025-250MG
APOSOR918025-1GEA 13860 CZK
APOSOR918025-1G APOSOR918025-250MG
3-Ethyl-3-oxetanecarboxaldehyde 95%
3-Ethyl-3-oxetanecarboxaldehyde
Vzorec: C₆H₁₀O₂
MW: 114.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18382018
CAS číslo: 98485-37-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR