4-Methylcyclohexylamine hydrochloride 95%
Dodavatel: Apollo Scientific
Synonyms:
4-Methylcyclohexanamine hydrochloride, Hexahydro-p-toluidine Hydrochloride
OR917951-25G
OR917951-5G
APOSOR917951-25GEA
12470
CZK
APOSOR917951-25G
APOSOR917951-5G
4-Methylcyclohexylamine hydrochloride 95%
4-Methylcyclohexylamine hydrochloride
Vzorec:
C₇H₁₅N MW: 149,66 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00051985 CAS číslo: 100959-19-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...