Tisk…
(S)-α-(2-Oxiranyl)anisole 95%
Dodavatel: Apollo Scientific
Synonyms: (S)-Glycidyl phenyl ether
OR917934-5G
APOSOR917934-5GEA 2730 CZK
APOSOR917934-5G
(S)-α-(2-Oxiranyl)anisole 95%
(S)-α-(2-Oxiranyl)anisole
Vzorec: C₉H₁₀O₂
MW: 150,1745 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06659026
CAS číslo: 71031-03-3