Tisk…

5-Ethynyl-2-methoxypyridine 95%

Dodavatel: Apollo Scientific

OR917903-1G OR917903-250MG
APOSOR917903-1GEA 5260 CZK
APOSOR917903-1G APOSOR917903-250MG
5-Ethynyl-2-methoxypyridine 95%
5-Ethynyl-2-methoxypyridine
Vzorec: C₈H₇NO
Storage Temperature: Ambient
MDL Number: MFCD10698135
CAS číslo: 663955-59-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR