2-(Methylthio)benzo[d]oxazole 95%
Dodavatel: Apollo Scientific
Synonyms:
2-(Methylthio)benzoxazole
OR917863-25G
APOSOR917863-25GEA
4140
CZK
APOSOR917863-25G
2-(Methylthio)benzo[d]oxazole 95%
2-(Methylthio)benzo[d]oxazole
Vzorec:
C₈H₇NOS MW: 165,2100 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00160027 CAS číslo: 13673-62-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...