Tisk…
1-Boc-1-methylhydrazine 96%
Dodavatel: Apollo Scientific
Synonyms: 1-tert-Butoxycarbonyl-1-methylhydrazine, 2-Methylcarbazic acid tert-Butyl ester, 1-Boc-1-methylhydrazine, tert-Butyl 2-Methylcarbazate
OR917775-100G
APOSOR917775-100GEA 2200 CZK
APOSOR917775-100G
1-Boc-1-methylhydrazine 96%
1-Boc-1-methylhydrazine
Vzorec: C₆H₁₄N₂O₂
MW: 146.1876 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05669700
CAS číslo: 21075-83-2