7-Methoxy-2H-chromene-3-carbaldehyde 95%
Dodavatel: Apollo Scientific
OR917741-1G
OR917741-250MG
APOSOR917741-1GEA
17990
CZK
APOSOR917741-1G
APOSOR917741-250MG
7-Methoxy-2H-chromene-3-carbaldehyde 95%
7-Methoxy-2H-chromene-3-carbaldehyde
Vzorec:
C₁₁H₁₀O₃ Storage Temperature: Ambient |
MDL Number:
MFCD01662419 CAS číslo: 57543-39-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...