2,3-Dihydroxybenzonitrile 95%
Dodavatel: Apollo Scientific
OR917737-5G
APOSOR917737-5GEA
2900
CZK
APOSOR917737-5G
2,3-Dihydroxybenzonitrile 95%
2,3-Dihydroxybenzonitrile
Vzorec:
C₇H₅NO₂ MW: 135.12 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02261930 CAS číslo: 67984-81-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...