3-Chloroquinoxalin-2-amine 95%
Dodavatel: Apollo Scientific
OR917720-1G
OR917720-250MG
APOSOR917720-1GEA
3610
CZK
APOSOR917720-1G
APOSOR917720-250MG
3-Chloroquinoxalin-2-amine 95%
3-Chloroquinoxalin-2-amine
Vzorec:
C₈H₆ClN₃ Storage Temperature: Ambient |
MDL Number:
MFCD00234398 CAS číslo: 34117-90-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...