Tisk…

4-Iodo-DL-phenylalanine 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: H-DL-Phe(4-I)-OH

OR917657-25G OR917657-5G
APOSOR917657-25GEA 22420 CZK
APOSOR917657-25G APOSOR917657-5G
4-Iodo-DL-phenylalanine 97%
4-Iodo-DL-phenylalanine
Vzorec: C₉H₁₀INO₂
MW: 291.1 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00063066
CAS číslo: 14173-41-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR