3-Bromo-1-methyl-1H-1,2,4-triazole 97%
Dodavatel: Apollo Scientific
OR917584-5G
APOSOR917584-5GEA
1950
CZK
APOSOR917584-5G
3-Bromo-1-methyl-1H-1,2,4-triazole 97%
3-Bromo-1-methyl-1H-1,2,4-triazole
Vzorec:
C3H4BrN3 MW: 161,9900 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11109755 CAS číslo: 56616-91-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...