Tisk…

3,7-Dimethyl-1H-indole-2-carbaldehyde 95%

Dodavatel: Apollo Scientific

OR917490-1G OR917490-250MG
APOSOR917490-1GEA 8270 CZK
APOSOR917490-1G APOSOR917490-250MG
3,7-Dimethyl-1H-indole-2-carbaldehyde 95%
3,7-Dimethyl-1H-indole-2-carbaldehyde
Vzorec: C₁₁H₁₁NO
Storage Temperature: Ambient
MDL Number: MFCD08361796
CAS číslo: 1463-72-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR