4-Chloro-2-(5-isoxazolyl)phenol 95%
Dodavatel: Apollo Scientific
OR917483-1G
OR917483-25G
OR917483-5G
APOSOR917483-1GEA
2900
CZK
APOSOR917483-1G
APOSOR917483-25G
APOSOR917483-5G
4-Chloro-2-(5-isoxazolyl)phenol 95%
4-Chloro-2-(5-isoxazolyl)phenol
Vzorec:
C₉H₆ClNO₂ MW: 195,61 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00218660 CAS číslo: 86176-56-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...