Cyclohexyl vinyl ether 95%
Dodavatel: Apollo Scientific
OR917443-100G
OR917443-1KG
OR917443-500G
APOSOR917443-100GEA
3540
CZK
APOSOR917443-100G
APOSOR917443-1KG
APOSOR917443-500G
Cyclohexyl vinyl ether 95%
Cyclohexyl vinyl ether
Vzorec:
C₈H₁₄O MW: 126,20 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00046358 CAS číslo: 2182-55-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...