Tisk…

4-Propylbenzaldehyde 96%

Dodavatel: Apollo Scientific

OR917431-1G OR917431-25G OR917431-5G
APOSOR917431-1GEA 695 CZK
APOSOR917431-1G APOSOR917431-25G APOSOR917431-5G
4-Propylbenzaldehyde 96%
4-Propylbenzaldehyde
Vzorec: C₁₀H₁₂O
Storage Temperature: Ambient
MDL Number: MFCD00041870
CAS číslo: 28785-06-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR