Tisk…

trans-Decahydroquinoline 98%

Dodavatel: Apollo Scientific

OR917395-100G OR917395-25G OR917395-5G
APOSOR917395-100GEA 22950 CZK
APOSOR917395-100G APOSOR917395-25G APOSOR917395-5G
trans-Decahydroquinoline 98%
trans-Decahydroquinoline
Vzorec: C₉H₁₇N
Storage Temperature: Ambient
MDL Number: MFCD00137755
CAS číslo: 767-92-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR