4-Bromo-2-furoic acid 95%
Dodavatel: Apollo Scientific
OR917328-1G
OR917328-250MG
APOSOR917328-1GEA
6680
CZK
APOSOR917328-1G
APOSOR917328-250MG
4-Bromo-2-furoic acid 95%
4-Bromo-2-furoic acid
Vzorec:
C₅H₃BrO₃ MW: 190,9800 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00092494 CAS číslo: 3439-02-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...