Tisk…

Methyl-2-(bromomethyl)-5-nitrobenzoate 96%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: Methyl 2-(bromomethyl)-5-nitrobenzoate

OR917272-1G OR917272-5G
APOSOR917272-1GEA 3380 CZK
APOSOR917272-1G APOSOR917272-5G
Methyl-2-(bromomethyl)-5-nitrobenzoate 96%
Methyl-2-(bromomethyl)-5-nitrobenzoate
Vzorec: C₉H₈BrNO₄
MW: 274,0681 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04114313
CAS číslo: 90725-68-1
UN: 3261
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR