N-(Carbobenzoxy)aminoacetonitrile 95%
Dodavatel: Apollo Scientific
Synonyms:
Benzyl (cyanomethyl)carbamate, Cyanomethylcarbamic Acid Benzyl Ester, Benzyl Cyanomethylcarbamate, N-Cbz-aminoacetonitrile
OR917260-25G
APOSOR917260-25GEA
6750
CZK
APOSOR917260-25G
N-(Carbobenzoxy)aminoacetonitrile 95%
N-(Carbobenzoxy)aminoacetonitrile
Vzorec:
C₁₀H₁₀N₂O₂ MW: 190,20 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00033175 CAS číslo: 3589-41-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...