Tisk…

3-Hydroxypyrazine-2-carboxamide 98%

Dodavatel: Apollo Scientific

Synonyms: T-1105

OR917252-100G
APOSOR917252-100GEA 3110 CZK
APOSOR917252-100G
3-Hydroxypyrazine-2-carboxamide 98%
3-Hydroxypyrazine-2-carboxamide
Vzorec: C₅H₅N₃O₂
MW: 139,114 g/mol
Bod tání: 270 °C
Storage Temperature: Ambient
MDL Number: MFCD00233977
CAS číslo: 55321-99-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR