Tisk…

2-Bromoethyl ethyl ether 97%

Dodavatel: Apollo Scientific

OR917179-100G OR917179-500G
APOSOR917179-100GEA 4770 CZK
APOSOR917179-100G APOSOR917179-500G
2-Bromoethyl ethyl ether 97%
2-Bromoethyl ethyl ether
Vzorec: C₄H₉BrO
MW: 153.02
Storage Temperature: Ambient
MDL Number: MFCD00000237
CAS číslo: 592-55-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR