Tisk…

S,S'-Diethyl dithiocarbonate 95%

Dodavatel: Apollo Scientific

OR916981-1G OR916981-5G
APOSOR916981-1GEA 9330 CZK
APOSOR916981-1G APOSOR916981-5G
S,S'-Diethyl dithiocarbonate 95%
S,S'-Diethyl dithiocarbonate
Vzorec: C₅H₁₀OS₂
Storage Temperature: Ambient
MDL Number: MFCD00144162
CAS číslo: 623-80-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR