2-Propylsulphanylpyrimidine-5-carbaldehyde 95%
Dodavatel: Apollo Scientific
OR916975-1G
OR916975-5G
APOSOR916975-1GEA
3940
CZK
APOSOR916975-1G
APOSOR916975-5G
2-Propylsulphanylpyrimidine-5-carbaldehyde 95%
2-Propylsulphanylpyrimidine-5-carbaldehyde
Vzorec:
C₈H₁₀N₂OS Storage Temperature: Ambient |
MDL Number:
MFCD07186427 CAS číslo: 876890-33-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...