Tisk…

Allyl-1-pyrrolidinecarbodithioate 95%

Dodavatel: Apollo Scientific

Synonyms: Allyl 1-Pyrrolidinecarbodithioate, 1-Pyrrolidinecarbodithioic acid Allyl Ester, 1-Pyrrolidinecarbodithioic Acid Allyl Ester, Pyrrolidinodithiocarbamic acid Allyl Ester, Pyrrolidinodithiocarbamic Acid Allyl Ester, Allyl Pyrrolidinodithiocarbamate

OR916805-1G OR916805-25G OR916805-5G
APOSOR916805-1GEA 3540 CZK
APOSOR916805-1G APOSOR916805-25G APOSOR916805-5G
Allyl-1-pyrrolidinecarbodithioate 95%
Allyl-1-pyrrolidinecarbodithioate
Vzorec: C₈H₁₃NS₂
MW: 187.32 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00135118
CAS číslo: 701-13-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR