Tisk…

Allyl butyl ether 97%

Dodavatel: Apollo Scientific

Synonyms: Allyl Butyl Ether, Butyl Allyl Ether, 1-(Allyloxy)butane

OR916709-25G
APOSOR916709-25GEA 2150 CZK
APOSOR916709-25G
Allyl butyl ether 97%
Allyl butyl ether
Vzorec: C₇H₁₄O
MW: 114,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00009460
CAS číslo: 3739-64-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR