Tisk…

2-(4-Methylpiperazin-1-yl)pyrimidin-4-amine 97%

Dodavatel: Apollo Scientific

OR916705-1G OR916705-250MG
APOSOR916705-1GEA 16960 CZK
APOSOR916705-1G APOSOR916705-250MG
2-(4-Methylpiperazin-1-yl)pyrimidin-4-amine 97%
2-(4-Methylpiperazin-1-yl)pyrimidin-4-amine
Vzorec: C₉H₁₅N₅
MW: 193,2500 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09999199
CAS číslo: 57005-71-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR