Tisk…

Butyl nitrate 95%

Dodavatel: Apollo Scientific

OR916679-100G OR916679-25G OR916679-5G OR916679-1G
APOSOR916679-100GEA 0 CZK
APOSOR916679-100G APOSOR916679-25G APOSOR916679-5G APOSOR916679-1G
Butyl nitrate 95%
Butyl nitrate
Vzorec: C₄H₉NO₃
MW: 119,12 g/mol
Storage Temperature: Ambient
CAS číslo: 928-45-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR