Tisk…

6-Hydroxy-1,4-benzodioxane 96%

Dodavatel: Apollo Scientific

Synonyms: 2,3-Dihydrobenzo[b][1,4]dioxin-6-ol

OR916527-1G OR916527-250MG OR916527-5G
APOSOR916527-1GEA 2570 CZK
APOSOR916527-1G APOSOR916527-250MG APOSOR916527-5G
6-Hydroxy-1,4-benzodioxane 96%
6-Hydroxy-1,4-benzodioxane
Vzorec: C₈H₈O₃
Storage Temperature: Ambient
MDL Number: MFCD06656571
CAS číslo: 10288-72-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR